Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01058789.hg.1 |
TC01003866.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01058789.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01058789.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000468172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000468172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000468172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000468172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000468172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000468172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000468172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000473144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000473144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000473144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000473144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000473144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000473144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000473144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000494899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000494899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
ENST00000494899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01058785.hg.1 |
TC01003866.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01058785.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01058785.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01058786.hg.1 |
TC01003866.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01058786.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01058786.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100231764.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100231764.hg.1 |
ENST00000494899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100231764.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100231764.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100231764.hg.1 |
ENST00000494899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100231764.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100231764.hg.1 |
TC0100017435.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100231761.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100231761.hg.1 |
ENST00000468172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100231761.hg.1 |
ENST00000494899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100231761.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100231761.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100231761.hg.1 |
ENST00000468172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100231761.hg.1 |
ENST00000494899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100231761.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100231761.hg.1 |
TC0100017435.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100231760.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100231760.hg.1 |
ENST00000468172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100231760.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100231760.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100231760.hg.1 |
ENST00000468172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100231760.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100231760.hg.1 |
TC0100017435.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000343410.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000468172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000468172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000468172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000468172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000468172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000468172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000468172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000473144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000473144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000473144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000473144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000473144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000473144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000473144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000494899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000494899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
ENST00000494899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017435.hg.1 |
NM_024746.3 |
Refseq |