Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST1H2AH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080596.2 | NP_542163.1 | 1 | 435 | 77 | |
GenBank mRNA | BC134366.1 | - | 1 | 388 | 40021708 | 77 |
BC148293.2 | - | 1 | 388 | 40021709 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04627640 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24817648 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04627650 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24817660 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24817659 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04627675 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24817641 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04627656 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24817662 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24817678 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04627644 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04627667 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24817644 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24817676 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24817657 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04627664 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24817658 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24817643 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24817665 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24817681 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24817652 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24817667 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24817645 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04627670 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04627663 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24817671 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 78604 | - | - |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 29617 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P81859 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P81859 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 20859 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P104729 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P104729 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 10928 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P81859 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P81859 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817644 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15796095 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817673 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817648 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817645 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15796099 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04627656 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817667 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04627667 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817655 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817657 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817636 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817652 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817643 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04627650 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817665 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04627645 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04627663 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817660 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817650 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04627640 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817641 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04627670 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817671 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817659 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817662 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817658 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817655 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04627667 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817667 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817671 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04627670 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817648 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04627656 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817652 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817665 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817659 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817660 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817650 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817658 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817636 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817673 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817645 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15796099 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817662 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817657 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04627650 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817643 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04627645 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817644 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817641 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04627663 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04627640 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15796095 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P81859 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P104729 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04627657 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04627653 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04627649 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04627659 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04627646 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04627666 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04627661 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04627654 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04627651 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04627645 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04627643 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04627760 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15796090 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04627763 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15796114 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04627762 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04627761 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P104729 | - | - |
Agilent Human Genome CGH Microarray 4x44K | A_14_P104729 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P81859 | - | - |
Custom Human Agilent Array 11k, Chromosome 6, Expression | 2163 | - | - |
Custom Human Agilent Array 11k, Chromosome 6, Expression | 9630 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 174451 | AK023683.1 | genbank_mrna_human |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 8086 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P104729 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04627649 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04627666 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04627662 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04627672 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15796097 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04627667 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04627651 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04627674 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15796090 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04627645 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04627653 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04627659 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04627678 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04627655 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15796099 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04627643 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P104729 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_080596_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8117546 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8117544 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8117545 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8117543 | - | - |
Affymetrix Human Genome U219 Array | 11736918_x_at | - | - |
Affymetrix Human Genome U219 Array | 11736918_PM_x_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0600014085.hg.1 | ENST00000377459.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014085.hg.1 | NM_080596.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014085.hg.1 | NM_080596.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011187.hg.1 | ENST00000627755.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06002092.hg.1 | TC06000225.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06002092.hg.1 | ENST00000377459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06002092.hg.1 | NM_080596.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014085.hg.1 | ENST00000377459.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014085.hg.1 | NM_080596.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014085.hg.1 | NM_080596.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600197776.hg.1 | NM_080596.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600197776.hg.1 | NM_080596.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600197776.hg.1 | TC0600014085.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600197777.hg.1 | ENST00000377459.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600197777.hg.1 | NM_080596.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600197777.hg.1 | ENST00000377459.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600197777.hg.1 | NM_080596.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600197777.hg.1 | TC0600014085.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600011187.hg.1 | ENST00000627755.1 | Ensembl |
Human Genome U133 Plus 2.0 Array | 214542_PM_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 208583_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 208583_PM_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 207156_at | - | - |
Human Genome U133 Plus 2.0 Array | 214542_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 214554_at | - | - |
Human Genome U133 Plus 2.0 Array | 207156_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 214554_PM_at | - | - |
Human Genome U133A 2.0 Array | 214542_x_at | - | - |
Human Genome U133A 2.0 Array | 207156_at | - | - |
Human Genome U133A 2.0 Array | 208583_x_at | - | - |
Human Genome U133A 2.0 Array | 207156_PM_at | - | - |
Human Genome U133A 2.0 Array | 214554_PM_at | - | - |
Human Genome U133A 2.0 Array | 214542_PM_x_at | - | - |
Human Genome U133A 2.0 Array | 214554_at | - | - |
Human Genome U133A 2.0 Array | 208583_PM_x_at | - | - |
Human Genome U95 Set | 285_g_at | - | - |
Human Genome U95 Set | 284_PM_at | - | - |
Human Genome U95 Set | 34157_PM_f_at | - | - |
Human Genome U95 Set | 31992_PM_f_at | - | - |
Human Genome U95 Set | 34157_f_at | - | - |
Human Genome U95 Set | 284_at | - | - |
Human Genome U95 Set | 285_PM_g_at | - | - |
Human Genome U95 Set | 34156_i_at | - | - |
Human Genome U95 Set | 31768_at | - | - |
Human Genome U95 Set | 34156_PM_i_at | - | - |
Human Genome U95 Set | 31992_f_at | - | - |
Human Genome U95 Set | 31768_PM_at | - | - |
Human Genome U133 Set | 214542_PM_x_at | - | - |
Human Genome U133 Set | 214554_at | - | - |
Human Genome U133 Set | 214554_PM_at | - | - |
Human Genome U133 Set | 208583_x_at | - | - |
Human Genome U133 Set | 207156_at | - | - |
Human Genome U133 Set | 207156_PM_at | - | - |
Human Genome U133 Set | 208583_PM_x_at | - | - |
Human Genome U133 Set | 214542_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031140 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031140 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_2188451 | NM_080596.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2188451 | BC093849.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2188451 | BC093851.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2188451 | BC134366.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2188451 | BC148293.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1789530 | - | - |
HumanHT-12 V4.0 expression beadchip | ILMN_2188451 | NM_080596.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2188451 | BC093849.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2188451 | BC093851.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2188451 | BC134366.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2188451 | BC148293.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2188451 | NM_080596.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2188451 | BC093849.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2188451 | BC093851.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2188451 | BC134366.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2188451 | BC148293.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2188451 | NM_080596.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2188451 | BC093849.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2188451 | BC093851.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2188451 | BC134366.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2188451 | BC148293.2 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_18105044-S | NM_080596.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_18105044-S | AK312232.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_18105044-S | BC093849.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_18105044-S | BC093851.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_18105044-S | BC134366.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_18090-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0000940451 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0000940451 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1789530 | - | - |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1789530 | - | - |
Illumina Sentrix Human-6 v2 Expression BeadChip | 2510403 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_18090 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2188451 | NM_080596.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2188451 | BC093849.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2188451 | BC093851.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2188451 | BC134366.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2188451 | BC148293.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2188451 | NM_080596.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2188451 | BC093849.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2188451 | BC093851.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2188451 | BC134366.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2188451 | BC148293.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2188451 | NM_080596.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2188451 | BC093849.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2188451 | BC093851.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2188451 | BC134366.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2188451 | BC148293.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1789530 | - | - |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2188451 | NM_080596.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2188451 | BC093849.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2188451 | BC093851.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2188451 | BC134366.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2188451 | BC148293.2 | human_transcriptome |