Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021148.hg.1 |
TC09001660.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021148.hg.1 |
ENST00000253018.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021148.hg.1 |
ENST00000372337.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021148.hg.1 |
ENST00000372338.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021148.hg.1 |
ENST00000448616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021148.hg.1 |
ENST00000454493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021148.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021148.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000372337.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000372337.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000372337.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000372337.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000372337.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000372337.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000372337.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000372337.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000372337.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000372338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000372338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000372338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000372338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000372338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000372338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000372338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000372338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000372338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000372338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000372338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000372338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000372338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000444139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000444139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000444139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000444139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000444139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000444139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000444139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000448616.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000448616.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000448616.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000448616.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000448616.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000448616.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000448616.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000448616.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000448616.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000448616.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000448616.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000448616.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000448616.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000451466.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000451466.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000451466.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000451466.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000451466.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000451466.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000451466.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000451466.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000463475.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000463475.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000463475.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000463475.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000486250.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000486250.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000486250.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000486250.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
ENST00000486250.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011755.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000372337.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000372337.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000372337.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000372337.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000372337.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000372337.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000372337.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000372337.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000372337.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000372338.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000372338.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000372338.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000372338.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000372338.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000372338.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000372338.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000372338.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000372338.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000372338.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000372338.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000372338.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000372338.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000444139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000444139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000444139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000444139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000444139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000444139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000444139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000448616.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000448616.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000448616.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000448616.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000448616.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000448616.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000448616.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000448616.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000448616.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000448616.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000448616.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000448616.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000448616.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000451466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000451466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000451466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000451466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000451466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000451466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000451466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000451466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000463475.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000463475.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000463475.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000463475.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000486250.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000486250.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000486250.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000486250.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
ENST00000486250.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011755.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900180324.hg.1 |
ENST00000372337.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180324.hg.1 |
ENST00000372338.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180324.hg.1 |
ENST00000444139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180324.hg.1 |
ENST00000448616.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180324.hg.1 |
ENST00000451466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180324.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900180324.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900180324.hg.1 |
ENST00000372337.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180324.hg.1 |
ENST00000372338.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180324.hg.1 |
ENST00000444139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180324.hg.1 |
ENST00000448616.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180324.hg.1 |
ENST00000451466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180324.hg.1 |
NM_001145106.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900180324.hg.1 |
NM_032843.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900180324.hg.1 |
TC0900011755.hg.1 |
Clariom D TC |