Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000476517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000476517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000476517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000476517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000476517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000613575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000613575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000613575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000613575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000613575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000613575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
ENST00000613575.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001325.hg.1 |
TC06000132.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001325.hg.1 |
ENST00000274695.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001325.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001325.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148597.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148597.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148597.hg.1 |
ENST00000613575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148597.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148597.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148597.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148597.hg.1 |
ENST00000613575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148597.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148597.hg.1 |
TC0600007143.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000274695.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000378610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000476517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000476517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000476517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000476517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000476517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000613575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000613575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000613575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000613575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000613575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000613575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
ENST00000613575.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007143.hg.1 |
NM_017774.3 |
Refseq |