Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025991.hg.1 |
ENST00000263093.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025991.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025991.hg.1 |
TC19001919.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000593745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000593745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000593745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000593745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000593745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000593745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000593745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000593745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000593745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000593745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000593745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000594683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000594683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000594683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000594683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000594786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000594786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000594786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000594786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000594786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000594786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000594786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000599700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000599700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000599700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000599700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000599700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000601997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000601997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000601997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
ENST00000601997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000593745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000593745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000593745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000593745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000593745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000593745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000593745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000593745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000593745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000593745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000593745.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000594683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000594683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000594683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000594683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000594786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000594786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000594786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000594786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000594786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000594786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000594786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000599700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000599700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000599700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000599700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000599700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000601997.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000601997.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000601997.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
ENST00000601997.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900012062.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900208566.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900208566.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900208566.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900208566.hg.1 |
TC1900012062.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900208566.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900208566.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900208566.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900208565.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900208565.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900208565.hg.1 |
NM_012254.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900208565.hg.1 |
TC1900012062.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900208565.hg.1 |
ENST00000263093.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900208565.hg.1 |
ENST00000601355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900208565.hg.1 |
NM_012254.2 |
Refseq |