Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000587561.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000587561.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000587561.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000587561.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000587561.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000587561.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000587918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000587918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000587918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000587940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000587940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000587940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000587940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000592562.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000592562.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000592562.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000592562.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000592562.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
ENST00000592562.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005675.hg.1 |
ENST00000251047.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005675.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005675.hg.1 |
TC18000547.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000587561.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000587561.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000587561.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000587561.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000587561.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000587561.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000587918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000587918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000587918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000587940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000587940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000587940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000587940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000592562.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000592562.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000592562.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000592562.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000592562.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
ENST00000592562.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008824.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800160748.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800160748.hg.1 |
NM_005570.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800160748.hg.1 |
TC1800008824.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800160748.hg.1 |
ENST00000251047.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800160748.hg.1 |
NM_005570.3 |
Refseq |