Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010849.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010849.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010849.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010849.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010849.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010849.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010849.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010849.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010849.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010849.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010849.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010849.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010849.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010849.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010849.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010849.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010849.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010849.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010849.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010849.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010849.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010849.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010849.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010849.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010849.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010849.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018492.hg.1 |
TC0X001376.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018492.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018492.hg.1 |
ENST00000535467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018492.hg.1 |
ENST00000536418.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018492.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018494.hg.1 |
TC0X001376.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018494.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018494.hg.1 |
ENST00000535467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018494.hg.1 |
ENST00000536418.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018494.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018498.hg.1 |
TC0X001376.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018498.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018498.hg.1 |
ENST00000536418.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018498.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173701.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173701.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173701.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173701.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173701.hg.1 |
TC0X00010849.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00010849.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010849.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010849.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010849.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010849.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010849.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010849.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010849.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010849.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010849.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010849.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010849.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010849.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010849.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010849.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010849.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010849.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010849.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010849.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010849.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010849.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010849.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010849.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010849.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010849.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010849.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173703.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173703.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173703.hg.1 |
TC0X00010849.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00173700.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173700.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173700.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173700.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173700.hg.1 |
TC0X00010849.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00173702.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173702.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173702.hg.1 |
ENST00000370828.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173702.hg.1 |
NM_001448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173702.hg.1 |
TC0X00010849.hg.1 |
Clariom D TC |