Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22015370.hg.1 |
ENST00000330651.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22015370.hg.1 |
ENST00000449719.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22015370.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22015370.hg.1 |
TC22000910.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000495277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000495277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000495277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000495277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000495277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
ENST00000495277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000495277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000495277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000495277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000495277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000495277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
ENST00000495277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009151.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200166280.hg.1 |
TC2200009151.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200166280.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200166280.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200166280.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200166280.hg.1 |
ENST00000495277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200166280.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200166280.hg.1 |
NR_110887.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200166280.hg.1 |
ENST00000330651.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200166280.hg.1 |
ENST00000395764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200166280.hg.1 |
ENST00000417877.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200166280.hg.1 |
ENST00000495277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200166280.hg.1 |
NM_002751.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200166280.hg.1 |
NR_110887.1 |
Refseq |