Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001045.hg.1 |
TC0X000079.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001045.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001045.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001045.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001045.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001041.hg.1 |
TC0X000079.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001041.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001041.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001041.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001044.hg.1 |
TC0X000079.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001044.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001044.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001044.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001040.hg.1 |
TC0X000079.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001040.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001040.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001043.hg.1 |
TC0X000079.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001043.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001043.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001043.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001043.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001042.hg.1 |
TC0X000079.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001042.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001042.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001042.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006697.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145867.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145867.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145867.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145867.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145867.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145867.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145867.hg.1 |
TC0X00006697.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00145868.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145868.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145868.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145868.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145868.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145868.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145868.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145868.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145868.hg.1 |
TC0X00006697.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00145866.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145866.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145866.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145866.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145866.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145866.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145866.hg.1 |
TC0X00006697.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00145864.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145864.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145864.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145864.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145864.hg.1 |
TC0X00006697.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00145865.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145865.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145865.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145865.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145865.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145865.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145865.hg.1 |
TC0X00006697.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00145869.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145869.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145869.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145869.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145869.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145869.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145869.hg.1 |
TC0X00006697.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00145870.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145870.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145870.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145870.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145870.hg.1 |
ENST00000380155.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145870.hg.1 |
ENST00000495743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145870.hg.1 |
NM_032796.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145870.hg.1 |
NR_033181.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145870.hg.1 |
TC0X00006697.hg.1 |
Clariom D TC |