Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017568.hg.1 |
TC09001408.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017568.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017568.hg.1 |
ENST00000375018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017568.hg.1 |
ENST00000375019.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017568.hg.1 |
ENST00000540940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017568.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017565.hg.1 |
TC09001408.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017565.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017565.hg.1 |
ENST00000375018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017565.hg.1 |
ENST00000375019.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017565.hg.1 |
ENST00000493393.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017565.hg.1 |
ENST00000540940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017565.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000444472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000444472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000444472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000444472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000444472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000444472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000444472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000444472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000444472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000466120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000466120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000466120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000466120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000471846.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000471846.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000471846.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000471846.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000471846.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000486778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000486778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000486778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
ENST00000486778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000444472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000444472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000444472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000444472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000444472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000444472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000444472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000444472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000444472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000466120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000466120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000466120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000466120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000471846.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000471846.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000471846.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000471846.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000471846.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000486778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000486778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000486778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
ENST00000486778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010980.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174837.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174837.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174837.hg.1 |
ENST00000444472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174837.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174837.hg.1 |
ENST00000353234.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174837.hg.1 |
ENST00000375019.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174837.hg.1 |
ENST00000444472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174837.hg.1 |
NM_173551.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174837.hg.1 |
TC0900010980.hg.1 |
Clariom D TC |