Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098616.2 | NP_001092086.1 | 2 - 3 | 558 | 99 | |
NM_001297717.1 | NP_001284646.1 | 2 - 3 | 558 | 99 | ||
NM_001297718.1 | NP_001284647.1 | 2 - 3 | 481 | 99 | ||
NM_001297720.1 | NP_001284649.1 | 2 - 3 | 558 | 99 | ||
NM_001297723.1 | NP_001284652.1 | 2 - 3 | 558 | 99 | ||
GenBank mRNA | AF109185.1 | - | 2 - 3 | 249 | 99 | |
AF529365.1 | - | 2 - 3 | 168 | 99 | ||
AK290583.1 | - | 2 - 3 | 330 | 99 | ||
AL540989.3 | - | 2 - 3 | 345 | 99 | ||
BC000152.2 | - | 2 - 3 | 367 | 2901006 | 99 | |
BC008306.1 | - | 2 - 3 | 294 | 3502049 | 99 | |
BC088353.1 | - | 2 - 3 | 378 | 6164281 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons |