Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FPR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002030.4 | NP_002021.3 | 2 | 753 | 149 | |
XM_011526687.2 | XP_011524989.1 | 2 | 644 | 149 | ||
GenBank mRNA | AK123030.1 | - | 3 | 696 | 149 | |
AK292541.1 | - | 3 | 742 | 149 | ||
BC059388.1 | - | 2 | 736 | 30332008 | 149 | |
BC069070.1 | - | 1 | 557 | 7216883 | 149 | |
BC069593.1 | - | 1 | 557 | 7262465 | 149 | |
BC069812.1 | - | 1 | 557 | 7262453 | 149 | |
M76673.1 | - | 1 | 557 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |