Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018299.hg.1 |
TC05001298.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018299.hg.1 |
ENST00000354209.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018299.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018300.hg.1 |
TC05001298.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018300.hg.1 |
ENST00000296800.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018300.hg.1 |
ENST00000354209.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018300.hg.1 |
ENST00000397128.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018300.hg.1 |
NM_006251.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018300.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000296800.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000296800.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000296800.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000296800.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000296800.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000296800.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000397006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000397006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000397006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000397128.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000397128.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000397128.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000397128.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000397128.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000397128.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000397128.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000397128.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000397128.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000397128.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000397128.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000397128.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000505783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000505783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000505783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000505783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000505783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000505783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000506652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000506652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000506652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000506652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000506652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000506652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000506652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000506652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000509874.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000509874.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000509874.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000509874.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000509874.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000511248.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000511248.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000511248.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000511248.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000511248.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000513152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
ENST00000513152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
NM_006251.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
NM_006251.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
NM_006251.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
NM_006251.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
NM_006251.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
NM_006251.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
NM_006251.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
NM_006251.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
NM_006251.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
NM_006251.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
NM_006251.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
NM_006251.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010580.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173837.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173837.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173837.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173837.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173837.hg.1 |
TC0500010580.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000296800.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000296800.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000296800.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000296800.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000296800.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000296800.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000397006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000397006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000397006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000397128.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000397128.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000397128.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000397128.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000397128.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000397128.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000397128.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000397128.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000397128.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000397128.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000397128.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000397128.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000505783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000505783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000505783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000505783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000505783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000505783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000506652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000506652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000506652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000506652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000506652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000506652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000506652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000506652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000509874.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000509874.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000509874.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000509874.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000509874.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000511248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000511248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000511248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000511248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000511248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000513152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
ENST00000513152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
NM_006251.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
NM_006251.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
NM_006251.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
NM_006251.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
NM_006251.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
NM_006251.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
NM_006251.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
NM_006251.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
NM_006251.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
NM_006251.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
NM_006251.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
NM_006251.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010580.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173838.hg.1 |
ENST00000296800.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173838.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173838.hg.1 |
ENST00000397128.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173838.hg.1 |
ENST00000506652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173838.hg.1 |
ENST00000509874.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173838.hg.1 |
NM_006251.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173838.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173838.hg.1 |
ENST00000296800.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173838.hg.1 |
ENST00000354209.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173838.hg.1 |
ENST00000397128.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173838.hg.1 |
ENST00000506652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173838.hg.1 |
ENST00000509874.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173838.hg.1 |
NM_006251.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173838.hg.1 |
NM_206907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173838.hg.1 |
TC0500010580.hg.1 |
Clariom D TC |