Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011237.hg.1 |
ENST00000303324.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011237.hg.1 |
ENST00000303324.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011237.hg.1 |
ENST00000303324.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011237.hg.1 |
NM_033057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011237.hg.1 |
NM_033057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011237.hg.1 |
NM_033057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016380.hg.1 |
TC06001434.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016380.hg.1 |
ENST00000303324.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016380.hg.1 |
NM_033057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016379.hg.1 |
TC06001434.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016379.hg.1 |
ENST00000303324.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016379.hg.1 |
NM_033057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011237.hg.1 |
ENST00000303324.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011237.hg.1 |
ENST00000303324.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011237.hg.1 |
ENST00000303324.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011237.hg.1 |
NM_033057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011237.hg.1 |
NM_033057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011237.hg.1 |
NM_033057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175754.hg.1 |
ENST00000303324.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175754.hg.1 |
NM_033057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175754.hg.1 |
ENST00000303324.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175754.hg.1 |
NM_033057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175754.hg.1 |
TC0600011237.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600175753.hg.1 |
ENST00000303324.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175753.hg.1 |
NM_033057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175753.hg.1 |
ENST00000303324.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175753.hg.1 |
NM_033057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175753.hg.1 |
TC0600011237.hg.1 |
Clariom D TC |