Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134336.1 | NP_001127808.1 | 18 - 19 | 1987 | 59 | |
NM_020704.2 | NP_065755.1 | 18 - 19 | 1987 | 59 | ||
XM_006716069.3 | XP_006716132.1 | 18 - 19 | 1987 | 59 | ||
XM_011516432.2 | XP_011514734.1 | 15 - 16 | 1635 | 59 | ||
XM_017012471.1 | XP_016867960.1 | 12 - 13 | 1310 | 59 | ||
XM_017012472.1 | XP_016867961.1 | 16 - 17 | 3452 | 59 | ||
GenBank mRNA | AB032996.1 | - | 18 - 19 | 1961 | 59 | |
AK290165.1 | - | 18 - 19 | 1987 | 59 | ||
AK302339.1 | - | 10 - 11 | 1192 | 59 | ||
BC019064.2 | - | 18 - 19 | 1974 | 4590587 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |