Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC29A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125343.1 | 9 - 10 | 1160 | 109 | ||
GenBank mRNA | AF401235.1 | - | 9 - 10 | 1168 | 109 | |
BC143335.1 | - | 9 - 10 | 980 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |