Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000265029.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000265029.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000265029.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000265029.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000265029.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000265029.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000265029.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000265029.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000265029.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000265029.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000265029.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000265029.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000382134.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000382134.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000382134.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000382134.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000382134.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000382134.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000382136.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000382136.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000382136.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000382136.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000382136.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000382136.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000382136.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000420570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000420570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000420570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000420570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000420570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000420570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000420570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000420570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000420570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000420570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000420570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000420570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000431018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000431018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000431018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000431018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000435961.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000435961.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000435961.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000435961.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000435961.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000435961.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000435961.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000435961.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000435961.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000435961.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000435961.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000435961.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000450521.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000450521.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000450521.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000450521.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000450521.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000450521.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000450521.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000450521.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000450521.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000488561.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
ENST00000488561.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_001308077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_001308077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_001308077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_001308077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_001308077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_001308077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_001308077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_001308077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_001308077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_001308077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_001308077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_001308079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_001308079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_001308079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_001308079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_001308079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_001308079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_001308079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_001308079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_001308079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_001308079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_014375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_014375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_014375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_014375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_014375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_014375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_014375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_014375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_014375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_014375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_014375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009773.hg.1 |
NM_014375.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018096.hg.1 |
TC03001021.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018096.hg.1 |
ENST00000265029.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018096.hg.1 |
ENST00000382134.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018096.hg.1 |
ENST00000382136.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018096.hg.1 |
ENST00000450521.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018096.hg.1 |
ENST00000488561.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018096.hg.1 |
ENST00000539949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018096.hg.1 |
NM_014375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000265029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000265029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000265029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000265029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000265029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000265029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000265029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000265029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000265029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000265029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000265029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000265029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000382134.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000382134.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000382134.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000382134.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000382134.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000382134.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000382136.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000382136.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000382136.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000382136.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000382136.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000382136.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000382136.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000420570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000420570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000420570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000420570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000420570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000420570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000420570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000420570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000420570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000420570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000420570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000420570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000431018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000431018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000431018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000431018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000435961.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000435961.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000435961.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000435961.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000435961.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000435961.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000435961.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000435961.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000435961.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000435961.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000435961.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000435961.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000450521.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000450521.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000450521.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000450521.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000450521.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000450521.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000450521.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000450521.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000450521.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000488561.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
ENST00000488561.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_001308077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_001308077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_001308077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_001308077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_001308077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_001308077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_001308077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_001308077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_001308077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_001308077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_001308077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_001308079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_001308079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_001308079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_001308079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_001308079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_001308079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_001308079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_001308079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_001308079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_001308079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_014375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_014375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_014375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_014375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_014375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_014375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_014375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_014375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_014375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_014375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_014375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009773.hg.1 |
NM_014375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171079.hg.1 |
ENST00000265029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171079.hg.1 |
ENST00000382134.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171079.hg.1 |
ENST00000382136.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171079.hg.1 |
ENST00000420570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171079.hg.1 |
ENST00000431018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171079.hg.1 |
ENST00000435961.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171079.hg.1 |
ENST00000450521.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171079.hg.1 |
ENST00000488561.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171079.hg.1 |
NM_001308077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171079.hg.1 |
NM_001308079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171079.hg.1 |
NM_014375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171079.hg.1 |
ENST00000265029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171079.hg.1 |
ENST00000382134.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171079.hg.1 |
ENST00000382136.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171079.hg.1 |
ENST00000420570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171079.hg.1 |
ENST00000431018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171079.hg.1 |
ENST00000435961.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171079.hg.1 |
ENST00000450521.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171079.hg.1 |
ENST00000488561.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171079.hg.1 |
NM_001308077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171079.hg.1 |
NM_001308079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171079.hg.1 |
NM_014375.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171079.hg.1 |
TC0300009773.hg.1 |
Clariom D TC |