Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007311.hg.1 |
ENST00000377575.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007311.hg.1 |
ENST00000377575.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007311.hg.1 |
ENST00000377575.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007311.hg.1 |
ENST00000377575.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007311.hg.1 |
ENST00000377575.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007311.hg.1 |
ENST00000477243.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007311.hg.1 |
ENST00000477243.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007311.hg.1 |
NM_006353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007311.hg.1 |
NM_006353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007311.hg.1 |
NM_006353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007311.hg.1 |
NM_006353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007311.hg.1 |
NM_006353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007311.hg.1 |
NM_006353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007311.hg.1 |
NM_006353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007311.hg.1 |
ENST00000377575.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007311.hg.1 |
ENST00000377575.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007311.hg.1 |
ENST00000377575.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007311.hg.1 |
ENST00000377575.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007311.hg.1 |
ENST00000377575.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007311.hg.1 |
ENST00000477243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007311.hg.1 |
ENST00000477243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007311.hg.1 |
NM_006353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007311.hg.1 |
NM_006353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007311.hg.1 |
NM_006353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007311.hg.1 |
NM_006353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007311.hg.1 |
NM_006353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007311.hg.1 |
NM_006353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007311.hg.1 |
NM_006353.2 |
Refseq |