Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014005.4 | NP_054724.1 | 1 | 5892 | 69 | |
GenBank mRNA | AB002343.1 | - | 1 | 5893 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon |