Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022551.hg.1 |
TC01001451.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022551.hg.1 |
ENST00000477875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022551.hg.1 |
NR_002925.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010481.hg.1 |
ENST00000477875.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010481.hg.1 |
ENST00000477875.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010481.hg.1 |
ENST00000477875.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010481.hg.1 |
ENST00000477875.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010481.hg.1 |
ENST00000477875.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010481.hg.1 |
ENST00000477875.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010481.hg.1 |
ENST00000477875.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010481.hg.1 |
ENST00000477875.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010481.hg.1 |
ENST00000488458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010481.hg.1 |
ENST00000488458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010481.hg.1 |
ENST00000488458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010481.hg.1 |
ENST00000488458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010481.hg.1 |
ENST00000488458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010481.hg.1 |
ENST00000488458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010481.hg.1 |
ENST00000488458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010481.hg.1 |
ENST00000488458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010481.hg.1 |
NR_002925.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010481.hg.1 |
NR_002925.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010481.hg.1 |
NR_002925.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010481.hg.1 |
NR_002925.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010481.hg.1 |
NR_002925.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010481.hg.1 |
NR_002925.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010481.hg.1 |
NR_002925.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022550.hg.1 |
TC01001451.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022550.hg.1 |
ENST00000477875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022550.hg.1 |
NR_002925.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177087.hg.1 |
ENST00000477875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177087.hg.1 |
ENST00000488458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177087.hg.1 |
NR_002925.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177087.hg.1 |
ENST00000477875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177087.hg.1 |
ENST00000488458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177087.hg.1 |
NR_002925.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177087.hg.1 |
TC0100010481.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100010481.hg.1 |
ENST00000477875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010481.hg.1 |
ENST00000477875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010481.hg.1 |
ENST00000477875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010481.hg.1 |
ENST00000477875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010481.hg.1 |
ENST00000477875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010481.hg.1 |
ENST00000477875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010481.hg.1 |
ENST00000477875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010481.hg.1 |
ENST00000477875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010481.hg.1 |
ENST00000488458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010481.hg.1 |
ENST00000488458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010481.hg.1 |
ENST00000488458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010481.hg.1 |
ENST00000488458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010481.hg.1 |
ENST00000488458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010481.hg.1 |
ENST00000488458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010481.hg.1 |
ENST00000488458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010481.hg.1 |
ENST00000488458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010481.hg.1 |
NR_002925.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010481.hg.1 |
NR_002925.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010481.hg.1 |
NR_002925.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010481.hg.1 |
NR_002925.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010481.hg.1 |
NR_002925.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010481.hg.1 |
NR_002925.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010481.hg.1 |
NR_002925.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177088.hg.1 |
ENST00000477875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177088.hg.1 |
ENST00000488458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177088.hg.1 |
NR_002925.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177088.hg.1 |
ENST00000477875.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177088.hg.1 |
ENST00000488458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177088.hg.1 |
NR_002925.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177088.hg.1 |
TC0100010481.hg.1 |
Clariom D TC |