Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001811.hg.1 |
ENST00000254854.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001811.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001811.hg.1 |
TC17000123.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
ENST00000574510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
ENST00000574510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
ENST00000574510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
ENST00000574510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700147826.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700147826.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700147826.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700147826.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700147826.hg.1 |
TC1700006784.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
ENST00000254854.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
ENST00000574510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
ENST00000574510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
ENST00000574510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
ENST00000574510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006784.hg.1 |
NM_000180.3 |
Refseq |