Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022416.hg.1 |
TC08001704.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022416.hg.1 |
ENST00000359228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022416.hg.1 |
NM_205545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022415.hg.1 |
TC08001704.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022415.hg.1 |
ENST00000359228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022415.hg.1 |
NM_205545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012086.hg.1 |
ENST00000359228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012086.hg.1 |
ENST00000359228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012086.hg.1 |
ENST00000359228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012086.hg.1 |
ENST00000359228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012086.hg.1 |
ENST00000359228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012086.hg.1 |
ENST00000359228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012086.hg.1 |
NM_205545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012086.hg.1 |
NM_205545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012086.hg.1 |
NM_205545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012086.hg.1 |
NM_205545.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012086.hg.1 |
NM_205545.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182187.hg.1 |
ENST00000359228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182187.hg.1 |
NM_205545.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182187.hg.1 |
ENST00000359228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182187.hg.1 |
NM_205545.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182187.hg.1 |
TC0800012086.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800182188.hg.1 |
ENST00000359228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182188.hg.1 |
NM_205545.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182188.hg.1 |
ENST00000359228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182188.hg.1 |
NM_205545.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182188.hg.1 |
TC0800012086.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800182186.hg.1 |
ENST00000359228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182186.hg.1 |
NM_205545.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182186.hg.1 |
ENST00000359228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182186.hg.1 |
NM_205545.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182186.hg.1 |
TC0800012086.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800012086.hg.1 |
ENST00000359228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012086.hg.1 |
ENST00000359228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012086.hg.1 |
ENST00000359228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012086.hg.1 |
ENST00000359228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012086.hg.1 |
ENST00000359228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012086.hg.1 |
ENST00000359228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012086.hg.1 |
NM_205545.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012086.hg.1 |
NM_205545.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012086.hg.1 |
NM_205545.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012086.hg.1 |
NM_205545.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012086.hg.1 |
NM_205545.2 |
Refseq |