Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007461.hg.1 |
TC02000487.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007461.hg.1 |
ENST00000272324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007461.hg.1 |
ENST00000393897.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007461.hg.1 |
ENST00000409471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007461.hg.1 |
NM_001008387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007461.hg.1 |
NM_198448.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000272324.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000272324.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000272324.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000272324.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000272324.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000272324.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000272324.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000272324.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000272324.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000272324.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000393897.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000393897.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000393897.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000393897.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000393897.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000393897.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000393897.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000393897.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000393897.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000409471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000409471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000409471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000409471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000409471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000409471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000409471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000490944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000490944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000490944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000490944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000490944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000498312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000498312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000498312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000498312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000498312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
ENST00000498312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
NM_001008387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
NM_001008387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
NM_001008387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
NM_001008387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
NM_001008387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
NM_001008387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
NM_001008387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
NM_001008387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
NM_001008387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
NM_001008387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
NM_001270040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
NM_001270040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
NM_001270040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
NM_001270040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
NM_001270040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
NM_001270040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
NM_001270040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
NM_001270040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
NM_001270040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
NM_198448.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
NM_198448.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
NM_198448.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
NM_198448.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
NM_198448.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
NM_198448.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
NM_198448.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
NM_198448.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008167.hg.1 |
NM_198448.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200156591.hg.1 |
ENST00000272324.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156591.hg.1 |
ENST00000393897.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156591.hg.1 |
ENST00000409471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156591.hg.1 |
ENST00000490944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156591.hg.1 |
ENST00000498312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156591.hg.1 |
NM_001008387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200156591.hg.1 |
NM_001270040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200156591.hg.1 |
NM_198448.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200156591.hg.1 |
ENST00000272324.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156591.hg.1 |
ENST00000393897.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156591.hg.1 |
ENST00000409471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156591.hg.1 |
ENST00000490944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156591.hg.1 |
ENST00000498312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156591.hg.1 |
NM_001008387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200156591.hg.1 |
NM_001270040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200156591.hg.1 |
NM_198448.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200156591.hg.1 |
TC0200008167.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000272324.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000272324.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000272324.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000272324.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000272324.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000272324.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000272324.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000272324.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000272324.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000272324.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000393897.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000393897.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000393897.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000393897.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000393897.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000393897.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000393897.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000393897.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000393897.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000409471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000409471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000409471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000409471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000409471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000409471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000409471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000490944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000490944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000490944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000490944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000490944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000498312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000498312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000498312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000498312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000498312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
ENST00000498312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
NM_001008387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
NM_001008387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
NM_001008387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
NM_001008387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
NM_001008387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
NM_001008387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
NM_001008387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
NM_001008387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
NM_001008387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
NM_001008387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
NM_001270040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
NM_001270040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
NM_001270040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
NM_001270040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
NM_001270040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
NM_001270040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
NM_001270040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
NM_001270040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
NM_001270040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
NM_198448.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
NM_198448.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
NM_198448.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
NM_198448.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
NM_198448.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
NM_198448.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
NM_198448.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
NM_198448.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008167.hg.1 |
NM_198448.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200156593.hg.1 |
ENST00000272324.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156593.hg.1 |
ENST00000393897.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156593.hg.1 |
ENST00000409471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156593.hg.1 |
ENST00000490944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156593.hg.1 |
ENST00000498312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156593.hg.1 |
NM_001008387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200156593.hg.1 |
NM_001270040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200156593.hg.1 |
NM_198448.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200156593.hg.1 |
ENST00000272324.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156593.hg.1 |
ENST00000393897.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156593.hg.1 |
ENST00000409471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156593.hg.1 |
ENST00000490944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156593.hg.1 |
ENST00000498312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200156593.hg.1 |
NM_001008387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200156593.hg.1 |
NM_001270040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200156593.hg.1 |
NM_198448.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200156593.hg.1 |
TC0200008167.hg.1 |
Clariom D TC |