Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022482.hg.1 |
TC08001709.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022482.hg.1 |
ENST00000323110.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022482.hg.1 |
NM_000498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012093.hg.1 |
ENST00000323110.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012093.hg.1 |
ENST00000323110.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012093.hg.1 |
ENST00000323110.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012093.hg.1 |
ENST00000323110.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012093.hg.1 |
ENST00000323110.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012093.hg.1 |
ENST00000323110.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012093.hg.1 |
ENST00000323110.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012093.hg.1 |
ENST00000323110.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012093.hg.1 |
ENST00000323110.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012093.hg.1 |
ENST00000323110.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012093.hg.1 |
NM_000498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012093.hg.1 |
NM_000498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012093.hg.1 |
NM_000498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012093.hg.1 |
NM_000498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012093.hg.1 |
NM_000498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012093.hg.1 |
NM_000498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012093.hg.1 |
NM_000498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012093.hg.1 |
NM_000498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012093.hg.1 |
NM_000498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012093.hg.1 |
NM_000498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012093.hg.1 |
ENST00000323110.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012093.hg.1 |
ENST00000323110.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012093.hg.1 |
ENST00000323110.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012093.hg.1 |
ENST00000323110.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012093.hg.1 |
ENST00000323110.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012093.hg.1 |
ENST00000323110.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012093.hg.1 |
ENST00000323110.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012093.hg.1 |
ENST00000323110.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012093.hg.1 |
ENST00000323110.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012093.hg.1 |
ENST00000323110.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012093.hg.1 |
NM_000498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012093.hg.1 |
NM_000498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012093.hg.1 |
NM_000498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012093.hg.1 |
NM_000498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012093.hg.1 |
NM_000498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012093.hg.1 |
NM_000498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012093.hg.1 |
NM_000498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012093.hg.1 |
NM_000498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012093.hg.1 |
NM_000498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012093.hg.1 |
NM_000498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182269.hg.1 |
ENST00000323110.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182269.hg.1 |
NM_000498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182269.hg.1 |
ENST00000323110.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182269.hg.1 |
NM_000498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182269.hg.1 |
TC0800012093.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800182268.hg.1 |
ENST00000323110.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182268.hg.1 |
NM_000498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182268.hg.1 |
ENST00000323110.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182268.hg.1 |
NM_000498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182268.hg.1 |
TC0800012093.hg.1 |
Clariom D TC |