Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000300179.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000300179.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000300179.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000300179.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000300179.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000300179.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000300179.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000300179.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000300179.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000300179.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000300179.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000300179.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000300179.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000454988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000454988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000454988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000454988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000454988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000454988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000454988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000454988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000489641.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000489641.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000489641.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000496985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000496985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000496985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000496985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
ENST00000496985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
NM_173564.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
NM_173564.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
NM_173564.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
NM_173564.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
NM_173564.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
NM_173564.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
NM_173564.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
NM_173564.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
NM_173564.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
NM_173564.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
NM_173564.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
NM_173564.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008548.hg.1 |
NM_173564.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000300179.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000300179.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000300179.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000300179.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000300179.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000300179.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000300179.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000300179.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000300179.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000300179.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000300179.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000300179.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000300179.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000454988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000454988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000454988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000454988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000454988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000454988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000454988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000454988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000489641.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000489641.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000489641.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000496985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000496985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000496985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000496985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
ENST00000496985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
NM_173564.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
NM_173564.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
NM_173564.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
NM_173564.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
NM_173564.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
NM_173564.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
NM_173564.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
NM_173564.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
NM_173564.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
NM_173564.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
NM_173564.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
NM_173564.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008548.hg.1 |
NM_173564.3 |
Refseq |