Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP6R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011530732.2 | XP_011529034.1 | 4 - 5 | 522 | 113 | |
XM_017029120.1 | XP_016884609.1 | 4 - 5 | 522 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons |