Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000471035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000471035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000471035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000471035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000471035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000482410.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000482410.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000482410.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000482410.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
ENST00000482410.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021870.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021870.hg.1 |
ENST00000369405.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021870.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021870.hg.1 |
TC10001667.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021869.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021869.hg.1 |
ENST00000369405.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021869.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021869.hg.1 |
TC10001667.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000471035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000471035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000471035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000471035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000471035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000482410.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000482410.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000482410.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000482410.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
ENST00000482410.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012595.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000188029.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000188029.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000188029.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000188029.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000188029.hg.1 |
ENST00000369404.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000188029.hg.1 |
ENST00000369405.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000188029.hg.1 |
NM_001303134.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000188029.hg.1 |
NM_022494.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000188029.hg.1 |
TC1000012595.hg.1 |
Clariom D TC |