Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022419.hg.1 |
TC09001742.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022419.hg.1 |
ENST00000316144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022419.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022419.hg.1 |
NM_203347.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
ENST00000316144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
ENST00000316144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
ENST00000316144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
ENST00000316144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
ENST00000316144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
ENST00000316144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
ENST00000316144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
ENST00000316144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
ENST00000316144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
ENST00000316144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
ENST00000316144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
ENST00000495223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
ENST00000495223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
ENST00000495223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
ENST00000495223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
NM_203347.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
NM_203347.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
NM_203347.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
NM_203347.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
NM_203347.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
NM_203347.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
NM_203347.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
NM_203347.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
NM_203347.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
NM_203347.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012003.hg.1 |
NM_203347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182386.hg.1 |
ENST00000316144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182386.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182386.hg.1 |
NM_203347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182386.hg.1 |
ENST00000316144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182386.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182386.hg.1 |
NM_203347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182386.hg.1 |
TC0900012003.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
ENST00000316144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
ENST00000316144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
ENST00000316144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
ENST00000316144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
ENST00000316144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
ENST00000316144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
ENST00000316144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
ENST00000316144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
ENST00000316144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
ENST00000316144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
ENST00000316144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
ENST00000482511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
ENST00000495223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
ENST00000495223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
ENST00000495223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
ENST00000495223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
NM_203347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
NM_203347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
NM_203347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
NM_203347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
NM_203347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
NM_203347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
NM_203347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
NM_203347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
NM_203347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
NM_203347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012003.hg.1 |
NM_203347.1 |
Refseq |