Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011255.hg.1 |
ENST00000289788.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011255.hg.1 |
ENST00000289788.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011255.hg.1 |
ENST00000289788.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011255.hg.1 |
ENST00000289788.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011255.hg.1 |
ENST00000289788.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011255.hg.1 |
ENST00000289788.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011255.hg.1 |
ENST00000289788.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011255.hg.1 |
ENST00000289788.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011255.hg.1 |
ENST00000481983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011255.hg.1 |
ENST00000481983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011255.hg.1 |
ENST00000481983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011255.hg.1 |
ENST00000481983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011255.hg.1 |
ENST00000481983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011255.hg.1 |
ENST00000481983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011255.hg.1 |
ENST00000486481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011255.hg.1 |
ENST00000486481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011255.hg.1 |
ENST00000486481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011255.hg.1 |
NM_001012455.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011255.hg.1 |
NM_001012455.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011255.hg.1 |
NM_001012455.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011255.hg.1 |
NM_001012455.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011255.hg.1 |
NM_001012455.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011255.hg.1 |
NM_001012455.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011255.hg.1 |
NM_001012455.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011255.hg.1 |
NM_001012455.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016480.hg.1 |
TC06001440.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016480.hg.1 |
ENST00000289788.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016480.hg.1 |
ENST00000486481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016480.hg.1 |
NM_001012455.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016478.hg.1 |
TC06001440.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016478.hg.1 |
ENST00000289788.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016478.hg.1 |
NM_001012455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175938.hg.1 |
ENST00000289788.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175938.hg.1 |
ENST00000481983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175938.hg.1 |
NM_001012455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175938.hg.1 |
ENST00000289788.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175938.hg.1 |
ENST00000481983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175938.hg.1 |
NM_001012455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175938.hg.1 |
TC0600011255.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600175939.hg.1 |
ENST00000289788.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175939.hg.1 |
ENST00000481983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175939.hg.1 |
NM_001012455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175939.hg.1 |
ENST00000289788.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175939.hg.1 |
ENST00000481983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175939.hg.1 |
NM_001012455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175939.hg.1 |
TC0600011255.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600175941.hg.1 |
ENST00000289788.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175941.hg.1 |
ENST00000481983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175941.hg.1 |
ENST00000486481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175941.hg.1 |
NM_001012455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175941.hg.1 |
ENST00000289788.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175941.hg.1 |
ENST00000481983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175941.hg.1 |
ENST00000486481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175941.hg.1 |
NM_001012455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175941.hg.1 |
TC0600011255.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600011255.hg.1 |
ENST00000289788.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011255.hg.1 |
ENST00000289788.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011255.hg.1 |
ENST00000289788.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011255.hg.1 |
ENST00000289788.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011255.hg.1 |
ENST00000289788.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011255.hg.1 |
ENST00000289788.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011255.hg.1 |
ENST00000289788.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011255.hg.1 |
ENST00000289788.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011255.hg.1 |
ENST00000481983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011255.hg.1 |
ENST00000481983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011255.hg.1 |
ENST00000481983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011255.hg.1 |
ENST00000481983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011255.hg.1 |
ENST00000481983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011255.hg.1 |
ENST00000481983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011255.hg.1 |
ENST00000486481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011255.hg.1 |
ENST00000486481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011255.hg.1 |
ENST00000486481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011255.hg.1 |
NM_001012455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011255.hg.1 |
NM_001012455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011255.hg.1 |
NM_001012455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011255.hg.1 |
NM_001012455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011255.hg.1 |
NM_001012455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011255.hg.1 |
NM_001012455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011255.hg.1 |
NM_001012455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011255.hg.1 |
NM_001012455.1 |
Refseq |