Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDH11X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168363.1 | NP_001161835.1 | 2 - 3 | 3077 | 142 | |
NM_032969.4 | NP_116751.1 | 2 - 3 | 3077 | 142 | ||
XM_011530913.2 | XP_011529215.1 | 6 - 7 | 3786 | 142 | ||
XM_017029418.1 | XP_016884907.1 | 6 - 7 | 3786 | 142 | ||
GenBank mRNA | AF332219.1 | - | 2 - 3 | 3878 | 142 | |
AY861434.1 | - | 2 - 3 | 3033 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Homo sapiens Inventoried Probe spans exons |