Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008171.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008171.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008171.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008171.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008171.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008171.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008171.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008171.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008171.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008171.hg.1 |
ENST00000432056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008171.hg.1 |
ENST00000432056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008171.hg.1 |
ENST00000432056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008171.hg.1 |
ENST00000432056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008171.hg.1 |
ENST00000612962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008171.hg.1 |
ENST00000613209.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008171.hg.1 |
NM_001282275.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008171.hg.1 |
NM_001282275.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008171.hg.1 |
NM_001282275.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008171.hg.1 |
NM_001282275.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008171.hg.1 |
NM_001282276.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008171.hg.1 |
NM_001282276.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008171.hg.1 |
NM_001282276.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008171.hg.1 |
NM_001282276.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011003.hg.1 |
ENST00000583892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011003.hg.1 |
NR_036150.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011003.hg.1 |
ENST00000583892.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011003.hg.1 |
NR_036150.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160375.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160375.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160375.hg.1 |
ENST00000613209.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160375.hg.1 |
NM_001282275.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160375.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160375.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160375.hg.1 |
ENST00000613209.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160375.hg.1 |
NM_001282275.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160375.hg.1 |
TC1700008171.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700160381.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160381.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160381.hg.1 |
ENST00000432056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160381.hg.1 |
NM_001282275.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160381.hg.1 |
NM_001282276.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160381.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160381.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160381.hg.1 |
ENST00000432056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160381.hg.1 |
NM_001282275.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160381.hg.1 |
NM_001282276.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160381.hg.1 |
TC1700008171.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700160380.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160380.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160380.hg.1 |
ENST00000432056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160380.hg.1 |
NM_001282275.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160380.hg.1 |
NM_001282276.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160380.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160380.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160380.hg.1 |
ENST00000432056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160380.hg.1 |
NM_001282275.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160380.hg.1 |
NM_001282276.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160380.hg.1 |
TC1700008171.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700008171.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008171.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008171.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008171.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008171.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008171.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008171.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008171.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008171.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008171.hg.1 |
ENST00000432056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008171.hg.1 |
ENST00000432056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008171.hg.1 |
ENST00000432056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008171.hg.1 |
ENST00000432056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008171.hg.1 |
ENST00000612962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008171.hg.1 |
ENST00000613209.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008171.hg.1 |
NM_001282275.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008171.hg.1 |
NM_001282275.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008171.hg.1 |
NM_001282275.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008171.hg.1 |
NM_001282275.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008171.hg.1 |
NM_001282276.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008171.hg.1 |
NM_001282276.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008171.hg.1 |
NM_001282276.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008171.hg.1 |
NM_001282276.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160382.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160382.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160382.hg.1 |
ENST00000432056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160382.hg.1 |
NM_001282275.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160382.hg.1 |
NM_001282276.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160382.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160382.hg.1 |
ENST00000422730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160382.hg.1 |
ENST00000432056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700160382.hg.1 |
NM_001282275.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160382.hg.1 |
NM_001282276.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700160382.hg.1 |
TC1700008171.hg.1 |
Clariom D TC |