Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCN5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB208866.1 | - | 18 | 6916 | 75 | |
AY038064.1 | - | 27 | 8290 | 75 | ||
EF092293.1 | - | 2 | 335 | 75 | ||
M77235.1 | - | 28 | 8396 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |