Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UFSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015072.3 | NP_001015072.2 | 1 | 948 | 83 | |
GenBank mRNA | AK292467.1 | - | 1 | 948 | 83 | |
BC017456.2 | - | 1 | 883 | 83 | ||
BC067904.1 | - | 1 | 911 | 5310615 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25902069 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25902071 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15996166 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05648562 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15996164 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15996165 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3327500 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3240053 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_1052 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_1051 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 47362 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 13123 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P18045207 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3327500 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3240053 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3240053 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3327500 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25902069 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25902071 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05648562 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15996165 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15996164 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05648562 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15996165 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15996164 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25902069 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25902071 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P05648562 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15996175 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15996176 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15996168 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15996167 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15996171 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15996170 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15996174 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15996172 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15996169 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15996173 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15996204 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15996169 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15996134 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05648515 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05648513 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05648585 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15996199 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15996198 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05648599 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15996166 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05648608 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15996174 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05648588 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05648607 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05648591 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05648511 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05648509 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15996202 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15996187 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05648602 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05648584 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05648596 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05648598 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05648603 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05648593 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15996193 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15996171 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15996176 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05648582 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_001015072_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8141646 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8141647 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8141644 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8141645 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8141643 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 240280_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11731345_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11731345_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0700012020.hg.1 | ENST00000388761.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012020.hg.1 | ENST00000388761.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012020.hg.1 | ENST00000388761.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012020.hg.1 | ENST00000388761.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012020.hg.1 | NM_001015072.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012020.hg.1 | NM_001015072.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012020.hg.1 | NM_001015072.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012020.hg.1 | NM_001015072.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700186403.hg.1 | ENST00000388761.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700186403.hg.1 | NM_001015072.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700186403.hg.1 | ENST00000388761.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700186403.hg.1 | NM_001015072.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700186403.hg.1 | TC0700012020.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700012020.hg.1 | ENST00000388761.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012020.hg.1 | ENST00000388761.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012020.hg.1 | ENST00000388761.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012020.hg.1 | ENST00000388761.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012020.hg.1 | NM_001015072.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012020.hg.1 | NM_001015072.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012020.hg.1 | NM_001015072.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012020.hg.1 | NM_001015072.3 | Refseq |
Human Genome U133 Plus 2.0 Array | 240280_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 240280_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_001015072_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240280_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240280_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240280_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240280_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240280_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240280_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240280_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240280_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240280_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240280_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240280_at2 | - | - |
Human Genome U95 Set | 67198_PM_at | - | - |
Human Genome U95 Set | 67198_at | - | - |
Human Genome U133 Set | 240280_at | - | - |
Human Genome U133 Set | 240280_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A033271 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A033271 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1771601 | BC067904.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1771601 | NM_001015072.3 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1771601 | AK292467.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1771601 | BC017456.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1771601 | BC067904.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1771601 | NM_001015072.3 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1771601 | AK292467.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1771601 | BC017456.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1771601 | BC067904.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1771601 | NM_001015072.3 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1771601 | AK292467.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1771601 | BC017456.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1771601 | BC067904.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1771601 | NM_001015072.3 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1771601 | AK292467.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1771601 | BC017456.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1771601 | BC067904.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1771601 | NM_001015072.3 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1771601 | AK292467.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1771601 | BC017456.2 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_27699-S | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1771601 | BC067904.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1771601 | NM_001015072.3 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1771601 | AK292467.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1771601 | BC017456.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1771601 | BC067904.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1771601 | NM_001015072.3 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1771601 | AK292467.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1771601 | BC017456.2 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_27699 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_27699 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1771601 | BC067904.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1771601 | NM_001015072.3 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1771601 | AK292467.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1771601 | BC017456.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1771601 | BC067904.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1771601 | NM_001015072.3 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1771601 | AK292467.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1771601 | BC017456.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1771601 | BC067904.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1771601 | NM_001015072.3 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1771601 | AK292467.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1771601 | BC017456.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1771601 | BC067904.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1771601 | NM_001015072.3 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1771601 | AK292467.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1771601 | BC017456.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1771601 | BC067904.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1771601 | NM_001015072.3 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1771601 | AK292467.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1771601 | BC017456.2 | human_transcriptome |