Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006489.hg.1 |
ENST00000298642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006489.hg.1 |
ENST00000298642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006489.hg.1 |
ENST00000298642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006489.hg.1 |
NM_001005501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000158.hg.1 |
ENST00000298642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000158.hg.1 |
NM_001005501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000158.hg.1 |
TC14000037.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1400006489.hg.1 |
ENST00000298642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006489.hg.1 |
ENST00000298642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006489.hg.1 |
ENST00000298642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006489.hg.1 |
NM_001005501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144312.hg.1 |
ENST00000298642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144312.hg.1 |
NM_001005501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144312.hg.1 |
ENST00000298642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144312.hg.1 |
NM_001005501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144312.hg.1 |
TC1400006489.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400144311.hg.1 |
ENST00000298642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144311.hg.1 |
ENST00000298642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144311.hg.1 |
TC1400006489.hg.1 |
Clariom D TC |