Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000605.3 | NP_000596.2 | 1 | 737 | 116 | |
GenBank mRNA | BC104163.1 | - | 1 | 714 | 40029595 | 116 |
BC104164.1 | - | 1 | 714 | 40029596 | 116 | |
M54886.1 | - | 1 | 729 | 117 | ||
V00544.1 | - | 1 | 729 | 117 | ||
V00548.1 | - | 1 | 651 | 116 | ||
V00549.1 | - | 1 | 729 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Human Cytokine Network Human Interferon Pathway HUMAN JAK-STAT PATHWAY Inventoried Human_Cytokine_Network Human_Interferon_Pathway Human_JAK-STAT_Pathway Both primers and probe map within a single exon |