Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000392948.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000392948.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000426275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000426275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000426275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000449108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000449108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000449108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000609585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000609585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000609585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000609585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000609585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000609879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000609879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000609879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015544.hg.1 |
ENST00000609879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015504.hg.1 |
ENST00000426275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015504.hg.1 |
ENST00000426275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015696.hg.1 |
ENST00000369167.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015696.hg.1 |
ENST00000369167.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015696.hg.1 |
ENST00000369167.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015696.hg.1 |
ENST00000369167.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015696.hg.1 |
ENST00000369167.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015696.hg.1 |
ENST00000369167.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015696.hg.1 |
ENST00000369167.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015696.hg.1 |
ENST00000420462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015696.hg.1 |
ENST00000420462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015696.hg.1 |
ENST00000420462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015696.hg.1 |
ENST00000469483.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015696.hg.1 |
ENST00000469483.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015696.hg.1 |
ENST00000469483.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015696.hg.1 |
ENST00000545683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015696.hg.1 |
ENST00000545683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015696.hg.1 |
ENST00000545683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015696.hg.1 |
ENST00000545683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015696.hg.1 |
ENST00000545683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015696.hg.1 |
ENST00000620458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015696.hg.1 |
ENST00000620458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015696.hg.1 |
NM_001024599.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015696.hg.1 |
NM_001024599.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015696.hg.1 |
NM_001024599.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015696.hg.1 |
NM_001161334.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015696.hg.1 |
NM_001161334.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015696.hg.1 |
NM_001161334.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015696.hg.1 |
NM_001161334.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015696.hg.1 |
NM_001161334.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100217535.hg.1 |
ENST00000369167.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100217535.hg.1 |
ENST00000369167.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100217535.hg.1 |
TC0100015696.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100217538.hg.1 |
ENST00000369167.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100217538.hg.1 |
NM_001024599.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100217538.hg.1 |
ENST00000369167.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100217538.hg.1 |
NM_001024599.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100217538.hg.1 |
TC0100015696.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100217537.hg.1 |
ENST00000369167.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100217537.hg.1 |
ENST00000369167.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100217537.hg.1 |
TC0100015696.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100015504.hg.1 |
ENST00000426275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015504.hg.1 |
ENST00000426275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000392948.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000392948.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000426275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000426275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000426275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000449108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000449108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000449108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000609585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000609585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000609585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000609585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000609585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000609879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000609879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000609879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015544.hg.1 |
ENST00000609879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100217536.hg.1 |
ENST00000369167.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100217536.hg.1 |
ENST00000420462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100217536.hg.1 |
ENST00000369167.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100217536.hg.1 |
ENST00000420462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100217536.hg.1 |
TC0100015696.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100015696.hg.1 |
ENST00000369167.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015696.hg.1 |
ENST00000369167.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015696.hg.1 |
ENST00000369167.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015696.hg.1 |
ENST00000369167.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015696.hg.1 |
ENST00000369167.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015696.hg.1 |
ENST00000369167.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015696.hg.1 |
ENST00000369167.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015696.hg.1 |
ENST00000420462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015696.hg.1 |
ENST00000420462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015696.hg.1 |
ENST00000420462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015696.hg.1 |
ENST00000469483.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015696.hg.1 |
ENST00000469483.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015696.hg.1 |
ENST00000469483.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015696.hg.1 |
ENST00000545683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015696.hg.1 |
ENST00000545683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015696.hg.1 |
ENST00000545683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015696.hg.1 |
ENST00000545683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015696.hg.1 |
ENST00000545683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015696.hg.1 |
ENST00000620458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015696.hg.1 |
ENST00000620458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015696.hg.1 |
NM_001024599.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015696.hg.1 |
NM_001024599.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015696.hg.1 |
NM_001024599.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015696.hg.1 |
NM_001161334.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015696.hg.1 |
NM_001161334.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015696.hg.1 |
NM_001161334.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015696.hg.1 |
NM_001161334.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015696.hg.1 |
NM_001161334.1 |
Refseq |