Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007380.hg.1 |
ENST00000225567.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007380.hg.1 |
ENST00000393456.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007380.hg.1 |
ENST00000415811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007380.hg.1 |
ENST00000439730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007380.hg.1 |
NM_001012511.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007380.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007380.hg.1 |
NM_054022.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007380.hg.1 |
TC17000603.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007381.hg.1 |
ENST00000415811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007381.hg.1 |
NM_001012511.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007381.hg.1 |
TC17000603.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000225567.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000225567.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000225567.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000225567.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000225567.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000225567.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000225567.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000225567.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000393456.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000393456.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000393456.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000393456.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000393456.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000393456.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000393456.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000393456.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000415811.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000415811.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000415811.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000415811.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000415811.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000415811.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000415811.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000570879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000570879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000570879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000570879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000570879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000570879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000571048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000571048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000571048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000571048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000571048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000571048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000571658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000571658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000571658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000575949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000575949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000575949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000575949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000575949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000575949.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000576910.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000576910.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000576910.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000576910.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000576910.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000576910.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000576910.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
ENST00000576910.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_001012511.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_001012511.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_001012511.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_001012511.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_001012511.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_001012511.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_001012511.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_054022.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_054022.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_054022.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_054022.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_054022.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_054022.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_054022.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_054022.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012271.hg.1 |
NM_054022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700202709.hg.1 |
ENST00000415811.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202709.hg.1 |
NM_001012511.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700202709.hg.1 |
ENST00000415811.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202709.hg.1 |
NM_001012511.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700202709.hg.1 |
TC1700012271.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700202708.hg.1 |
ENST00000225567.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202708.hg.1 |
ENST00000393456.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202708.hg.1 |
ENST00000415811.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202708.hg.1 |
ENST00000570879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202708.hg.1 |
NM_001012511.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700202708.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700202708.hg.1 |
NM_054022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700202708.hg.1 |
ENST00000225567.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202708.hg.1 |
ENST00000393456.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202708.hg.1 |
ENST00000415811.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202708.hg.1 |
ENST00000570879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202708.hg.1 |
NM_001012511.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700202708.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700202708.hg.1 |
NM_054022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700202708.hg.1 |
TC1700012271.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000225567.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000225567.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000225567.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000225567.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000225567.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000225567.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000225567.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000225567.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000393456.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000393456.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000393456.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000393456.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000393456.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000393456.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000393456.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000393456.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000415811.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000415811.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000415811.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000415811.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000415811.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000415811.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000415811.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000570879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000570879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000570879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000570879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000570879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000570879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000571048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000571048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000571048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000571048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000571048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000571048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000571658.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000571658.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000571658.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000575949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000575949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000575949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000575949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000575949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000575949.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000576910.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000576910.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000576910.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000576910.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000576910.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000576910.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000576910.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
ENST00000576910.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_001012511.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_001012511.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_001012511.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_001012511.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_001012511.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_001012511.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_001012511.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_004287.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_054022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_054022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_054022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_054022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_054022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_054022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_054022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_054022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012271.hg.1 |
NM_054022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700202710.hg.1 |
ENST00000415811.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202710.hg.1 |
NM_001012511.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700202710.hg.1 |
ENST00000415811.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202710.hg.1 |
NM_001012511.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700202710.hg.1 |
TC1700012271.hg.1 |
Clariom D TC |