Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000327564.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000327564.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000327564.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000327564.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000327564.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000327564.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000327564.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000327564.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000327564.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000327564.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000327564.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000327564.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000327564.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000327564.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000327564.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000327564.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000327564.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000341671.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000341671.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000341671.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000341671.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000341671.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000341671.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000341671.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000341671.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000341671.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000341671.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000341671.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000341671.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000341671.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000341671.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000341671.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000394713.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000394713.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000394713.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000394713.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000394713.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000394713.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000394713.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000394713.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000394713.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000394713.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000394713.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000394713.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000394713.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000394713.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000394713.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000578118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000578118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000578118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000578118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000578118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000578674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000578674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000578674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000578674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000578674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000578674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000578674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000578674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000578674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000578674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000578674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000578674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000578674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000579094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000579094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000579094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000579094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000579094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000579094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000579766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000579766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000579766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000579766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000579766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000580303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000580303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000580303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000580303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000580303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000580303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000580871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000580871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000580871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000580871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000580871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000580871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000580871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000580871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000581086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000581086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000581086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000581086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000581786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000581786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000581786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000581786.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000582117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000582117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000582117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000582117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000582815.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000582815.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000582815.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000582815.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000582815.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000582815.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000583204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000583204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000583204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000583204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000583204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000583204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000583204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000583204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000583204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000583204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000583204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000583758.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000583758.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000583758.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000583758.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000583758.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000583758.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000583758.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000583758.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000584368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000584368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000584368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000584368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000584368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000584368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000584368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000584368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000584368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000584368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000584368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
ENST00000584368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
NM_052888.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
NM_052888.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
NM_052888.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
NM_052888.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
NM_052888.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
NM_052888.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
NM_052888.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
NM_052888.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
NM_052888.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
NM_052888.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
NM_052888.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
NM_052888.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
NM_052888.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
NM_052888.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012228.hg.1 |
NM_052888.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000585915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000585915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000585915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000585915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000590010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000590019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000590019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000590019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000590019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011551.hg.1 |
ENST00000590019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000585915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000585915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000585915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000585915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000586022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000590010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000590019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000590019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000590019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000590019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011551.hg.1 |
ENST00000590019.1 |
Ensembl |