Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNORD12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_003695.1 | 1 | 48 | 98 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BG402259.1 | - | 157 | Chr. - - [-] on Build | 98 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ZNFX1 antisense RNA 1 | ZFAS1 |
C20orf199, HSUP1, HSUP2, NCRNA00275, ZNFX1-AS1 |
Sense | Chr. 20 - 49278178 - 49289260 on NCBI Build GRCh38 | |
Downstream |
small nucleolar RNA, C/D box 12 |
SNORD12 |
HBII-99, MIR1259, MIRN1259 |
268.0 | Sense | Chr. 20 - 49280683 - 49280772 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |