Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | TSNAX-DISC1 readthrough (NMD candidate) | TSNAX-DISC1 | Anti-sense | Chr. 1 - 231528653 - 232041272 on NCBI Build GRCh38 | ||
Downstream | DISC1 intronic transcript 1 | DISC1-IT1 | 100.9kb | Anti-sense | Chr. 1 - 231925834 - 231945205 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |