Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST7-AS2 | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF400040.1 | - | LIT1999 ST7OT2 |
2 - 3 | 253 | 84 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 6132 | MIR6132 | hsa-mir-6132 | -60.5kb | Anti-sense | Chr. 7 - 117020211 - 117020319 on NCBI Build GRCh38 |
Downstream | ST7 overlapping transcript 3 | ST7-OT3 |
NCRNA00026, ST7, ST7OT3 |
38.4kb | Anti-sense | Chr. 7 - 117182681 - 117209937 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |