Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PWAR5 | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_022008.1 | 1 | 2538 | 147 | |||
GenBank mRNA | AF019618.1 | - | LIT1110 PAR5 |
1 | 2348 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 377931 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P921610 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P921610 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 15947 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 42245 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P20185646 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_18_P12146754 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P20185653 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P20185644 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P02994316 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P921610 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_24_P921610 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30022934 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543083 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30022966 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30022957 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543085 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30022959 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30022962 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543092 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543087 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543088 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30022955 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30022933 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30022956 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30022958 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30022968 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30022963 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30022964 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30022960 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30022966 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543088 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30022959 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543085 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30022964 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30022960 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543092 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30022934 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30022962 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30022933 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30022963 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30022957 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543083 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30022968 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30022955 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30022958 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30022956 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543087 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P921610 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P921610 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543185 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543202 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543165 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543102 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543149 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543137 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543068 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543070 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543150 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543122 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543156 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543176 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543166 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543190 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543119 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543124 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543191 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543168 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543088 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543062 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543140 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543154 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543061 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543135 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543144 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543057 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543075 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543142 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543183 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543193 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543085 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543200 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543133 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543177 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543114 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543052 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543123 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543110 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543083 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543058 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543147 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543163 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543131 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543113 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543170 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543157 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543196 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16760168 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543066 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543116 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543073 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543111 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543091 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543064 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543047 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543161 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543188 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543195 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543179 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543126 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543199 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543079 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543173 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543159 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543117 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543054 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543152 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NR_022008_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7981944 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7981943 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 214834_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR15000598.hg.1 | ENST00000459332.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15000598.hg.1 | NR_001292.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15000598.hg.1 | NR_022008.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15000598.hg.1 | TC15000045.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR15000600.hg.1 | NR_022008.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15000600.hg.1 | TC15000045.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1500006569.hg.1 | ENST00000386683.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006569.hg.1 | ENST00000459332.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006569.hg.1 | ENST00000605533.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006569.hg.1 | ENST00000605533.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006569.hg.1 | ENST00000605533.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006569.hg.1 | ENST00000624480.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006569.hg.1 | ENST00000624480.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006569.hg.1 | ENST00000624480.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006569.hg.1 | ENST00000624480.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006569.hg.1 | ENST00000624480.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006569.hg.1 | ENST00000624480.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006569.hg.1 | ENST00000624480.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006569.hg.1 | NR_001292.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006569.hg.1 | NR_001294.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006569.hg.1 | NR_022008.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006569.hg.1 | NR_022008.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006569.hg.1 | NR_022008.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006569.hg.1 | NR_022008.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006569.hg.1 | NR_022008.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006569.hg.1 | NR_022008.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006569.hg.1 | NR_022008.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006569.hg.1 | NR_022008.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500145715.hg.1 | ENST00000624480.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500145715.hg.1 | NR_022008.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500145715.hg.1 | ENST00000624480.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500145715.hg.1 | NR_022008.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500145715.hg.1 | TC1500006569.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500145717.hg.1 | ENST00000624480.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500145717.hg.1 | NR_022008.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500145717.hg.1 | ENST00000624480.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500145717.hg.1 | NR_022008.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500145717.hg.1 | TC1500006569.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1500006569.hg.1 | ENST00000386683.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006569.hg.1 | ENST00000459332.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006569.hg.1 | ENST00000605533.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006569.hg.1 | ENST00000605533.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006569.hg.1 | ENST00000605533.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006569.hg.1 | ENST00000624480.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006569.hg.1 | ENST00000624480.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006569.hg.1 | ENST00000624480.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006569.hg.1 | ENST00000624480.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006569.hg.1 | ENST00000624480.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006569.hg.1 | ENST00000624480.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006569.hg.1 | ENST00000624480.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006569.hg.1 | NR_001292.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006569.hg.1 | NR_001294.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006569.hg.1 | NR_022008.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006569.hg.1 | NR_022008.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006569.hg.1 | NR_022008.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006569.hg.1 | NR_022008.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006569.hg.1 | NR_022008.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006569.hg.1 | NR_022008.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006569.hg.1 | NR_022008.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006569.hg.1 | NR_022008.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500145716.hg.1 | ENST00000624480.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500145716.hg.1 | NR_022008.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500145716.hg.1 | ENST00000624480.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500145716.hg.1 | NR_022008.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500145716.hg.1 | TC1500006569.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500145713.hg.1 | ENST00000459332.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500145713.hg.1 | ENST00000624480.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500145713.hg.1 | NR_001292.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500145713.hg.1 | NR_022008.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500145713.hg.1 | ENST00000459332.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500145713.hg.1 | ENST00000624480.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500145713.hg.1 | NR_001292.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500145713.hg.1 | NR_022008.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500145713.hg.1 | TC1500006569.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 214834_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 214834_at | - | - |
Human Genome U133A 2.0 Array | 214834_PM_at | - | - |
Human Genome U133A 2.0 Array | 214834_at | - | - |
Human Genome U95 Set | 35490_at | - | - |
Human Genome U95 Set | 35490_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214834_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214834_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214834_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214834_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214834_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214834_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214834_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214834_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214834_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214834_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214834_at8 | - | - |
Human Genome U133 Set | 214834_at | - | - |
Human Genome U133 Set | 214834_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A052655 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A052655 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3241803 | AF019618.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3241803 | NR_022008.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3241803 | AF019618.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3241803 | NR_022008.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3241803 | AF019618.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3241803 | NR_022008.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3241803 | AF019618.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3241803 | NR_022008.1 | human_transcriptome |