Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
testis-specific transcript, Y-linked 8B (non-protein coding) |
TTTY8B |
NCRNA00217, TTTY8 |
Sense | Chr. Y - 6471031 - 6473630 on NCBI Build GRCh38 | |
Downstream |
amelogenin, Y-linked |
AMELY |
AMGL, AMGY |
392.9kb | Sense | Chr. Y - 6865918 - 6874056 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Homo sapiens Probe spans exons |