Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
histone cluster 1, H1c |
HIST1H1C |
H1.2, H1C, H1F2, H1s-1 |
-29.8kb | Anti-sense | Chr. 6 - 26055740 - 26056471 on NCBI Build GRCh38 |
Downstream | hemochromatosis | HFE |
HFE1, HH, HLA-H, MVCD7, TFQTL2 |
858.0 | Sense | Chr. 6 - 26087281 - 26096216 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |