Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010739.hg.1 |
TC09002885.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010739.hg.1 |
ENST00000247665.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010739.hg.1 |
ENST00000371661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010739.hg.1 |
ENST00000492540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010739.hg.1 |
ENST00000545326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010739.hg.1 |
NM_001135861.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010739.hg.1 |
NM_014172.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010742.hg.1 |
TC09002885.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010742.hg.1 |
ENST00000247665.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010742.hg.1 |
ENST00000371661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010742.hg.1 |
ENST00000492540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010742.hg.1 |
ENST00000545326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010742.hg.1 |
NM_001135861.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010742.hg.1 |
NM_014172.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000247665.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000247665.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000247665.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000247665.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000247665.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000247665.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000462205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000462205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000462205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000462205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000462205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000462205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000463215.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000463215.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000463215.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000463215.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000463215.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000492540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000492540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000492540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000492540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000492540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000492540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000492540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000492540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000497413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000497413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000497413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000497413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000497413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
ENST00000497413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_001135861.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_001135861.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_001135861.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_001135861.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_001135861.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_001135861.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_001135861.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_001135861.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_001287342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_001287342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_001287342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_001287342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_001287342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_001287342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_001287342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_001287343.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_001287343.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_001287343.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_001287343.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_001287343.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_001287343.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_001287343.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_001287343.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_014172.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_014172.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_014172.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_014172.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_014172.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_014172.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NM_014172.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NR_109807.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NR_109807.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NR_109807.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NR_109807.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NR_109807.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NR_109807.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NR_109807.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NR_109807.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NR_109807.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NR_109808.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NR_109808.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NR_109808.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NR_109808.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NR_109808.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NR_109808.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012192.hg.1 |
NR_109808.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010736.hg.1 |
TC09002885.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010736.hg.1 |
ENST00000545326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010736.hg.1 |
NM_001135861.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010736.hg.1 |
NM_014172.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010744.hg.1 |
TC09002885.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010744.hg.1 |
ENST00000247665.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010744.hg.1 |
ENST00000371661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010744.hg.1 |
ENST00000492540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010744.hg.1 |
ENST00000545326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010744.hg.1 |
NM_001135861.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010744.hg.1 |
NM_014172.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000247665.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000247665.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000247665.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000247665.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000247665.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000247665.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000462205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000462205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000462205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000462205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000462205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000462205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000463215.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000463215.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000463215.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000463215.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000463215.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000492540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000492540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000492540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000492540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000492540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000492540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000492540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000492540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000497413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000497413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000497413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000497413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000497413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
ENST00000497413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_001135861.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_001135861.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_001135861.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_001135861.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_001135861.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_001135861.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_001135861.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_001135861.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_001287342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_001287342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_001287342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_001287342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_001287342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_001287342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_001287342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_001287343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_001287343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_001287343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_001287343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_001287343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_001287343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_001287343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_001287343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_014172.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_014172.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_014172.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_014172.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_014172.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_014172.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NM_014172.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NR_109807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NR_109807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NR_109807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NR_109807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NR_109807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NR_109807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NR_109807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NR_109807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NR_109807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NR_109808.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NR_109808.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NR_109808.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NR_109808.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NR_109808.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NR_109808.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012192.hg.1 |
NR_109808.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900185344.hg.1 |
ENST00000247665.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185344.hg.1 |
ENST00000462205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185344.hg.1 |
ENST00000463215.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185344.hg.1 |
ENST00000492540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185344.hg.1 |
ENST00000497413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185344.hg.1 |
NM_001135861.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900185344.hg.1 |
NM_001287342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900185344.hg.1 |
NM_001287343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900185344.hg.1 |
NM_014172.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900185344.hg.1 |
NR_109807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900185344.hg.1 |
NR_109808.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900185344.hg.1 |
ENST00000247665.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185344.hg.1 |
ENST00000462205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185344.hg.1 |
ENST00000463215.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185344.hg.1 |
ENST00000492540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185344.hg.1 |
ENST00000497413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185344.hg.1 |
NM_001135861.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900185344.hg.1 |
NM_001287342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900185344.hg.1 |
NM_001287343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900185344.hg.1 |
NM_014172.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900185344.hg.1 |
NR_109807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900185344.hg.1 |
NR_109808.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900185344.hg.1 |
TC0900012192.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900185345.hg.1 |
ENST00000492540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185345.hg.1 |
NR_109807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900185345.hg.1 |
ENST00000492540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185345.hg.1 |
NR_109807.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900185345.hg.1 |
TC0900012192.hg.1 |
Clariom D TC |