Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
ENST00000521412.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
ENST00000521412.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
ENST00000521412.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
ENST00000521412.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
ENST00000521412.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
ENST00000521412.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
ENST00000521412.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
ENST00000521412.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
ENST00000521412.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
ENST00000521412.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
ENST00000521412.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
NR_024035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
NR_024035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
NR_024035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
NR_024035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
NR_024035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
NR_024035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
NR_024035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
NR_024035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013029.hg.1 |
NR_024035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05029610.hg.1 |
TC05002127.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05029610.hg.1 |
ENST00000503486.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05029610.hg.1 |
NR_024035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500191180.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500191180.hg.1 |
ENST00000521412.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500191180.hg.1 |
NR_024035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500191180.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500191180.hg.1 |
ENST00000521412.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500191180.hg.1 |
NR_024035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500191180.hg.1 |
TC0500013029.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
ENST00000521412.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
ENST00000521412.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
ENST00000521412.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
ENST00000521412.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
ENST00000521412.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
ENST00000521412.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
ENST00000521412.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
ENST00000521412.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
ENST00000521412.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
ENST00000521412.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
ENST00000521412.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
NR_024035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
NR_024035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
NR_024035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
NR_024035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
NR_024035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
NR_024035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
NR_024035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
NR_024035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013029.hg.1 |
NR_024035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500191171.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500191171.hg.1 |
NR_024035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500191171.hg.1 |
ENST00000503486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500191171.hg.1 |
NR_024035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500191171.hg.1 |
TC0500013029.hg.1 |
Clariom D TC |