Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05012366.hg.1 |
TC05000852.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05012366.hg.1 |
ENST00000297109.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05012366.hg.1 |
ENST00000426761.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05012366.hg.1 |
ENST00000440364.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05012366.hg.1 |
ENST00000523198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05012366.hg.1 |
NM_001131062.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05012366.hg.1 |
NM_001131063.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05012366.hg.1 |
NM_024632.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05012366.hg.1 |
NR_024084.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500164128.hg.1 |
ENST00000297109.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164128.hg.1 |
ENST00000426761.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164128.hg.1 |
ENST00000440364.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164128.hg.1 |
ENST00000517926.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164128.hg.1 |
ENST00000520159.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164128.hg.1 |
ENST00000520731.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164128.hg.1 |
ENST00000523029.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164128.hg.1 |
ENST00000523198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164128.hg.1 |
NM_001131062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500164128.hg.1 |
NM_001131063.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500164128.hg.1 |
NM_024632.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500164128.hg.1 |
NR_024084.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500164128.hg.1 |
ENST00000297109.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164128.hg.1 |
ENST00000426761.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164128.hg.1 |
ENST00000440364.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164128.hg.1 |
ENST00000517926.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164128.hg.1 |
ENST00000520159.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164128.hg.1 |
ENST00000520731.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164128.hg.1 |
ENST00000523029.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164128.hg.1 |
ENST00000523198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164128.hg.1 |
NM_001131062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500164128.hg.1 |
NM_001131063.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500164128.hg.1 |
NM_024632.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500164128.hg.1 |
NR_024084.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500164128.hg.1 |
TC0500009161.hg.1 |
Clariom D TC |