Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000547164.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000547164.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000547164.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200160465.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200160465.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200160465.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200160465.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200160465.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200160465.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200160465.hg.1 |
TC1200008223.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000547164.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000547164.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000547164.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |