Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009506.hg.1 |
ENST00000517299.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009506.hg.1 |
ENST00000517299.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009506.hg.1 |
ENST00000517299.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009506.hg.1 |
ENST00000517299.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009506.hg.1 |
ENST00000519897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009506.hg.1 |
ENST00000519897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009506.hg.1 |
ENST00000519897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009506.hg.1 |
ENST00000519897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009506.hg.1 |
NR_027108.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009506.hg.1 |
NR_027108.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009506.hg.1 |
NR_027108.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009506.hg.1 |
NR_027108.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009506.hg.1 |
NR_027109.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009506.hg.1 |
NR_027109.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009506.hg.1 |
NR_027109.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009506.hg.1 |
NR_027109.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05013987.hg.1 |
TC05000964.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05013987.hg.1 |
ENST00000519897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05013987.hg.1 |
NR_027109.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009506.hg.1 |
ENST00000517299.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009506.hg.1 |
ENST00000517299.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009506.hg.1 |
ENST00000517299.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009506.hg.1 |
ENST00000517299.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009506.hg.1 |
ENST00000519897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009506.hg.1 |
ENST00000519897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009506.hg.1 |
ENST00000519897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009506.hg.1 |
ENST00000519897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009506.hg.1 |
NR_027108.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009506.hg.1 |
NR_027108.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009506.hg.1 |
NR_027108.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009506.hg.1 |
NR_027108.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009506.hg.1 |
NR_027109.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009506.hg.1 |
NR_027109.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009506.hg.1 |
NR_027109.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009506.hg.1 |
NR_027109.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500166506.hg.1 |
ENST00000519897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166506.hg.1 |
NR_027109.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500166506.hg.1 |
ENST00000519897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166506.hg.1 |
NR_027109.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500166506.hg.1 |
TC0500009506.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500166507.hg.1 |
ENST00000519897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166507.hg.1 |
NR_027109.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500166507.hg.1 |
ENST00000519897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166507.hg.1 |
NR_027109.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500166507.hg.1 |
TC0500009506.hg.1 |
Clariom D TC |