Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | epithelial membrane protein 1 | EMP1 |
CL-20, EMP-1, TMP |
-3.9kb | Sense | Chr. 12 - 13196668 - 13216774 on NCBI Build GRCh38 |
Downstream | long intergenic non-protein coding RNA 1559 | LINC01559 | C12orf36 | 148.8kb | Anti-sense | Chr. 12 - 13370671 - 13376745 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |